Maleic acid 99%

Supplier: Thermo Scientific Chemicals
Danger

12523-0010 12523-0051 12523-0250 12523-1000
TS12523-0010EA 91 USD
TS12523-0010 TS12523-0051 TS12523-0250 TS12523-1000
Maleic acid 99%
Maleic acid
Formula: C₄H₄O₄
MW: 116.07 g/mol
Melting Pt: 134…138 °C
Flash Pt: 127 °C
Storage Temperature: Ambient
MDL Number: MFCD00063177
CAS Number: 110-16-7
EINECS: 203-742-5
UN: 3261
Merck Index: 13,05726

Order Now

Specification Test Results

Appearance White Crystalline powder or granules and/or chunks
Infrared spectrum Conforms
Melting point 130 to 138 °C
Water ≤2 %
HPLC ≥98.5 %
Titration with NaOH 98.5 to 101.5 % (on dry substance)
Heavy metals (as Pb) ≤10 ppm

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR