Mineral oil (petroleum) for IR spectrophotometry, suitable for nujol mull preparations

Supplier: Thermo Scientific Chemicals
Danger

12402-0010 12402-0025 12402-0060 12402-0100
TS12402-0010EA 59 USD
TS12402-0010 TS12402-0025 TS12402-0060 TS12402-0100
Mineral oil (petroleum) for IR spectrophotometry, suitable for nujol mull preparations
Mineral oil (petroleum)
Density: 0.851 g/cm³
Flash Pt: 185 °C
Storage Temperature: Ambient
MDL Number: MFCD00131611
CAS Number: 8042-47-5

Order Now

Specification Test Results

Appearance Clear colorless Oily liquid
Infrared spectrum Conforms
Refractive index 1.4620 to 1.4730 (20 °C, 589 nm)
Density (15°C) 830 to 865 kg/l
Viscosity 13 to 19 mm²/s (40°C)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR