4-Oxopimelic acid 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 4-Oxoheptanedioic acid, 4-Ketopimelic acid

12288-0050 12288-0250 12288-1000
TS12288-0050EA 78 USD
TS12288-0050 TS12288-0250 TS12288-1000
4-Oxopimelic acid 98%
4-Oxopimelic acid
Formula: C₇H₁₀O₅
MW: 174.15 g/mol
Melting Pt: 142…144 °C
Storage Temperature: Ambient
MDL Number: MFCD00002798
CAS Number: 502-50-1
EINECS: 207-941-8

Order Now

Specification Test Results

Appearance White to yellow Powder or crystals and/or chunks
Infrared spectrum Conforms
Melting point 141 to 145 °C
Titration with NaOH ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR