Itaconic acid 99+%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 2-Methylenesuccinic acid, Methylenesuccinic acid

12281-0010 12281-0051 12281-2500
TS12281-0010EA 49 USD
TS12281-0010 TS12281-0051 TS12281-2500
Itaconic acid 99+%
Itaconic acid
Formula: C₅H₆O₄
MW: 130.1 g/mol
Melting Pt: 165…168 °C
Density: 1.57 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00004260
CAS Number: 97-65-4
EINECS: 202-599-6
Merck Index: 13,05264

Order Now

Specification Test Results

Appearance White to light beige Crystalline powder or crystals
Infrared spectrum Conforms
Melting point 165 to 168 °C
Water ≤0.5 %
Sulfated ash ≤0.1 %
Titration with NaOH ≥99.0 %
Sulfate (SO4) ≤300 ppm
Heavy metals (as Pb) ≤20 ppm

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR