1,3-Indandione 97%

Supplier: Thermo Scientific Chemicals
Warning

12209-0050 12209-0250 12209-1000
TS12209-0050EA 45 USD
TS12209-0050 TS12209-0250 TS12209-1000
1,3-Indandione 97%
1,3-Indandione
Formula: C₉H₆O₂
Melting Pt: 129…132 °C
Storage Temperature: Ambient
MDL Number: MFCD00003779
CAS Number: 606-23-5
EINECS: 210-109-7

Order Now

Specification Test Results

Appearance Light yellow to beige to golden Powder
Infrared spectrum Conforms
Melting point 124 to 131 °C
Titration with NaOH ≥96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR