4-Hydroxy-4-methyl-2-pentanone 99%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Diacetone alcohol

12148-0010 12148-0025 12148-0100 12148-2500
TS12148-0010EA 70 USD
TS12148-0010 TS12148-0025 TS12148-0100 TS12148-2500
4-Hydroxy-4-methyl-2-pentanone 99%
4-Hydroxy-4-methyl-2-pentanone
Formula: (CH₃)₂C(OH)CH₂COCH₃
MW: 116.16 g/mol
Boiling Pt: 166
Melting Pt: -44
Density: 0.9385 g/cm³
Flash Pt: 58 °C
Storage Temperature: Ambient
MDL Number: MFCD00004471
CAS Number: 123-42-2
EINECS: 204-626-7
UN: 1148
ADR: 3 III
REACH: 01-2119473975-21
Merck Index: 13,02983

Order Now

Specification Test Results

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4220 to 1.4250 (20 °C, 589 nm)
GC ≥98.5 %
Color scale ≤10 APHA


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR