5-(Hydroxymethyl)-2-furaldehyde 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 5-Hydroxymethyl-2-furancarbaldehyde, 5-(hydroxymethyl)furan-2-carbaldehyde, 5-(Hydroxymethyl)furfural, 5-Hydroxymethylfurfural

12146-0010 12146-0050 12146-0250
TS12146-0010EA 58 USD
TS12146-0010 TS12146-0050 TS12146-0250
5-(Hydroxymethyl)-2-furaldehyde 98%
5-(Hydroxymethyl)-2-furaldehyde
Formula: C₆H₆O₃
MW: 126.11 g/mol
Boiling Pt: 114…116 °C (1 mmHg)
Melting Pt: 30…34 °C
Flash Pt: 79 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00003234
CAS Number: 67-47-0
EINECS: 200-654-9
Merck Index: 13,04857

Order Now

Specification Test Results

Appearance Colorless to white to yellow to orange to brown Liquid or crystalline powder and/or chunks or fused solid
Infrared spectrum Conforms
Melting point 28 to 34 °C
GC ≥99.50 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR