cis-3-Hexen-1-ol 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
cis-Hex-3-en-1-ol
12077-0100
12077-0500
TS12077-0100EA
48
USD
TS12077-0100
TS12077-0500
cis-3-Hexen-1-ol 98%
cis-3-Hexen-1-ol
Formula:
C₆H₁₂O MW: 100.16 g/mol Boiling Pt: 156…157 °C (760 mmHg) Melting Pt: –60 °C Density: 0.848 g/cm³ Flash Pt: 59 °C Storage Temperature: Ambient |
MDL Number:
MFCD00063217 CAS Number: 928-96-1 EINECS: 213-192-8 UN: 1987 ADR: 3,III Merck Index: 13,04719 |
Specification Test Results
Appearance | Clear colorless to light yellow Liquid |
Infrared spectrum | Conforms |
Refractive index | 1.4380 to 1.4410 (20 °C, 589 nm) |
Density | (20°C) 0.845 to 0.853 g/ml |
GC | ≥97.5 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...