1-Hexadecanol 96%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Cetyl alcohol, Palmityl alcohol

12048-0010 12048-0025 12048-0100
TS12048-0010EA 60 USD
TS12048-0010 TS12048-0025 TS12048-0100
1-Hexadecanol 96%
1-Hexadecanol
Formula: H₃C(CH₂)₁₄CH₂OH
MW: 242.45 g/mol
Boiling Pt: 344 °C (760 mmHg)
Melting Pt: 47…50 °C
Flash Pt: >157 °C
Storage Temperature: Ambient
MDL Number: MFCD00004760
CAS Number: 36653-82-4
EINECS: 253-149-0
Merck Index: 13,02037

Order Now

Specification Test Results

Appearance White to off-white Powder or flakes or pellets
Infrared spectrum Conforms
GC ≥95 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR