3-Heptanone 98%

Supplier: Thermo Scientific Chemicals
Warning

12038-0010 12038-0025 12038-2500
TS12038-0010EA 255 USD
TS12038-0010 TS12038-0025 TS12038-2500
3-Heptanone 98%
3-Heptanone
Formula: C₇H₁₄O
MW: 114.19 g/mol
Boiling Pt: 146…149 °C (760 mmHg)
Melting Pt: –39 °C
Density: 0.81 g/cm³
Flash Pt: 41 °C
Storage Temperature: Ambient
MDL Number: MFCD00009483
CAS Number: 106-35-4
EINECS: 203-388-1
UN: 1224
ADR: 3,III

Order Now

Specification Test Results

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4075 to 1.4095 (20 °C, 589 nm)
GC ≥97.5 %
NMR (1H-NMR) Consistent with structure

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR