7-Isopropyl-1,4-dimethylazulene 99%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Guaiazulene

12020-0100 12020-0250
TS12020-0100EA 63 USD
TS12020-0100 TS12020-0250
7-Isopropyl-1,4-dimethylazulene 99%
7-Isopropyl-1,4-dimethylazulene
Formula: C₁₅H₁₈
MW: 198.31 g/mol
Boiling Pt: 153 °C (7Torr)
Melting Pt: 29…31 °C
Density: 0.97 g/cm³
Flash Pt: >110 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00003811
CAS Number: 489-84-9
EINECS: 207-701-2
Merck Index: 12,04581

Order Now

Specification Test Results

Appearance Dark blue Low melting crystalline solid
Infrared spectrum Conforms
GC ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR