Glycyrrhetic acid 98+%

Supplier: Thermo Scientific Chemicals

Synonyms: 18 beta-Glycyrrhetic acid

12015-0050 12015-0250
TS12015-0050EA 66 USD
TS12015-0050 TS12015-0250
Glycyrrhetic acid 98+%
Glycyrrhetic acid
Formula: C₃₀H₄₆O₄
MW: 470.69 g/mol
Melting Pt: 292…295 °C
Storage Temperature: Ambient
MDL Number: MFCD00003706
CAS Number: 471-53-4

Order Now

Specification Test Results

Appearance White to off-white Crystalline powder
Infrared spectrum Conforms
Loss on drying ≤1 % (105°C, 4 h)
Sulfated ash ≤0.2 %
Related substances ≤2.0 % (HPLC)
Specific optical rotation +145° to +154° (20°C, 589 nm) (c=1, dioxane) (on dried substance)
Titration with NaOH 98.0 to 102.0 % (on dried substance)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR