Glutaric acid 99%

Supplier: Thermo Scientific Chemicals
Warning

11996-0250 11996-1000 11996-5000
TS11996-0250EA 32 USD
TS11996-0250 TS11996-1000 TS11996-5000
Glutaric acid 99%
Glutaric acid
Formula: C₅H₈O₄
Boiling Pt: 302 °C (1013 hPa)
Melting Pt: 92…99 °C
Storage Temperature: Ambient
MDL Number: MFCD00004410
CAS Number: 110-94-1
EINECS: 203-817-2
Merck Index: 13,04486

Order Now

Specification Test Results

Appearance White to off-white Crystalline powder or crystals
Infrared spectrum Conforms
Melting point 95 to 99 °C
Titration with NaOH 98.5 to 101.5 %
NMR (1H-NMR) Consistent with structure


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR