1-Fluoro-2-nitrobenzene 99%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: o-Fluoronitrobenzene, 2-Fluoronitrobenzene

11948-0100 11948-0500 11948-2500
TS11948-0100EA 24 USD
TS11948-0100 TS11948-0500 TS11948-2500
1-Fluoro-2-nitrobenzene 99%
1-Fluoro-2-nitrobenzene
Formula: C₆H₄FNO₂
MW: 141.1 g/mol
Boiling Pt: 215 °C (760 mmHg)
Melting Pt: 9…6 °C
Density: 1.335 g/cm³
Flash Pt: 94 °C
Storage Temperature: Ambient
MDL Number: MFCD00007048
CAS Number: 1493-27-2
EINECS: 216-088-0
UN: 2810
ADR: 6.1,II

Order Now

Specification Test Results

Appearance Clear yellow to brown Liquid
Infrared spectrum Conforms
Refractive index 1.5307 to 1.5327 (20 °C, 589 nm)
GC ≥98.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR