4'-Fluoroacetophenone 99%

Supplier: Thermo Scientific Chemicals
Warning

11926-0250 11926-1000 11926-5000
TS11926-0250EA 33 USD
TS11926-0250 TS11926-1000 TS11926-5000
4'-Fluoroacetophenone 99%
4'-Fluoroacetophenone
Formula: C₈H₇FO
Boiling Pt: 196 °C (760 mmHg)
Melting Pt: 4 °C
Density: 1.13 g/cm³
Flash Pt: 71 °C
Storage Temperature: Ambient
MDL Number: MFCD00000354
CAS Number: 403-42-9
EINECS: 206-960-9

Order Now

Specification Test Results

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.5090 to 1.5120 (20 °C, 589 nm)
GC ≥98.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR