1-Ethynylcyclohexanol 99+%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: 1-Ethynyl-1-cyclohexanol

11910-0025 11910-1000 11910-5000
TS11910-0025EA 675 USD
TS11910-0025 TS11910-1000 TS11910-5000
1-Ethynylcyclohexanol 99+%
1-Ethynylcyclohexanol
Formula: C₈H₁₂O
MW: 124.18 g/mol
Boiling Pt: 180 °C (760 mmHg)
Melting Pt: 30…32 °C
Density: 0.9763 g/cm³
Flash Pt: 73 °C
Storage Temperature: Ambient
MDL Number: MFCD00003858
CAS Number: 78-27-3
EINECS: 241-385-7
UN: 2811
ADR: 6.1,III

Order Now

Specification Test Results

Appearance Clear colorless to light yellow Liquid after melting
Infrared spectrum Conforms
Refractive index 1.4810 to 1.4840 (20 °C, 589 nm)
GC ≥99.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR