2-Ethylcaproic acid 99%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
2-Ethylhexanoic acid
11852-0011
11852-0025
11852-0100
11852-5000
TS11852-0011EA
62
USD
TS11852-0011
TS11852-0025
TS11852-0100
TS11852-5000
2-Ethylcaproic acid 99%
2-Ethylcaproic acid
Formula:
C₈H₁₆O₂ Boiling Pt: 228 °C (760 mmHg) Melting Pt: –59 °C Density: 0.906 g/cm³ Flash Pt: 114 °C Storage Temperature: Ambient |
MDL Number:
MFCD00002675 CAS Number: 149-57-5 EINECS: 205-743-6 UN: 3265 ADR: 8,III |
Specification Test Results
Appearance | Clear liquid |
Infrared spectrum | Conforms |
Refractive index | 1.4240 to 1.4260 (20 °C, 589 nm) |
Water | ≤0.1 % (K.F.) |
GC | ≥98.5 % |
Color scale | ≤15 APHA |
Iron (Fe) | ≤5 ppm |
Heavy metals (as Pb) | ≤5 ppm |
Insoluble matter | ≤0.005 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...