Ethyl-2-oxocyclohexanecarboxylate 95%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
Ethyl 2-oxocyclohexanecarboxylate
11833-0050
11833-0250
11833-1000
TS11833-0050EA
34
USD
TS11833-0050
TS11833-0250
TS11833-1000
Ethyl-2-oxocyclohexanecarboxylate 95%
Ethyl-2-oxocyclohexanecarboxylate
Formula:
C₉H₁₄O₃ MW: 170.21 g/mol Boiling Pt: 106 °C (11 mmHg) Density: 1.06 g/cm³ Flash Pt: 85 °C Storage Temperature: Ambient |
MDL Number:
MFCD00001631 CAS Number: 1655-07-8 EINECS: 216-731-5 |
Specification Test Results
Appearance | Clear colorless to light yellow Liquid |
Infrared spectrum | Conforms |
Refractive index | 1.4775 to 1.4795 (20 °C, 589 nm) |
GC | ≥94 % |
Molybdenum (Mo) | ≤10 ppm |
Heavy metals & Fe (as Pb) | ≤0.001 % |
Titrable free base | ≤0.02 meq/g |
Insoluble matter | ≤0.01 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...