Ethyl butyrylacetate 98%

Supplier: Thermo Scientific Chemicals

Synonyms: Ethyl Butyrylacetate, Ethyl 3-oxohexanoate

11819-0250 11819-1000
TS11819-0250EA 28 USD
TS11819-0250 TS11819-1000
Ethyl butyrylacetate 98%
Ethyl butyrylacetate
Formula: C₈H₁₄O₃
Boiling Pt: 104 °C (22 mmHg)
Density: 1 g/cm³
Flash Pt: 78 °C
Storage Temperature: Ambient
MDL Number: MFCD00009401
CAS Number: 3249-68-1
EINECS: 221-835-9

Order Now

Specification Test Results

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4260 to 1.4290 (20 °C, 589 nm)
GC ≥97.5 %
Insoluble matter ≤0.01 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR