Ethyl bromopyruvate 80-85%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Bromopyruvic acid ethyl ester, Ethyl 3-bromo-2-oxopropanoate, 3-Bromo-2-oxopropionic acid ethyl ester

11815-0250 11815-1000
TS11815-0250EA 108 USD
TS11815-0250 TS11815-1000
Ethyl bromopyruvate 80-85%
Ethyl bromopyruvate
Formula: BrH₂CC(O)COOCH₂CH₃
Boiling Pt: 100…102 °C (15 mmHg)
Density: 1.55 g/cm³
Flash Pt: 98 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00000204
CAS Number: 70-23-5
EINECS: 200-729-6
UN: 3265
ADR: 8,III

Order Now

Specification Test Results

Appearance Clear yellow to pink-red Liquid
Infrared spectrum Conforms
GC ≥80 %
Phosphate (PO4) ≤10 ppm
Sulfate (SO4) ≤30 ppm
Sodium carbonate (Na2CO3) ≤1.0 %
Calcium (Ca) ≤50 ppm
Iron (Fe) ≤10 ppm
Magnesium (Mg) ≤20 ppm
Mercury (Hg) ≤0.1 ppm
Nickel (Ni) ≤10 ppm
Potassium (K) ≤200 ppm
Heavy metals (as Ag) ≤20 ppm
Nitrogen compounds (as N) ≤0.001 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR