Dipentaerythritol 85+%, Technical Grade

Supplier: Thermo Scientific Chemicals

Synonyms: 2,2'-(Oxybis(methylene))bis(2-(hydroxymethyl)propane-1,3-diol)

11712-0250 11712-5000
TS11712-0250EA 30 USD
TS11712-0250 TS11712-5000
Dipentaerythritol 85+%, Technical Grade
Dipentaerythritol
Formula: C₁₀H₂₂O₇
MW: 254.28 g/mol
Melting Pt: 215…225 °C
Flash Pt: >150 °C
Storage Temperature: Ambient
MDL Number: MFCD00004691
CAS Number: 126-58-9
EINECS: 204-794-1

Order Now

Specification Test Results

Appearance White Powder
Infrared spectrum Conforms
Melting point 215 to 225 °C
GC ≥85.0 %
Hydroxyl value 1125 to 1325 mg KOH/g
Residue after evaporation ≤0.003 %
Substance darkened by H2SO4 passes test
Titrable acid ≤0.0009 meq/g

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR