Dimethyl glutarate 98%

Supplier: Thermo Scientific Chemicals

Synonyms: 1,5-Pentanedioic acid dimethyl ester, Glutaric acid dimethyl ester

11623-1000 11623-5000
TS11623-1000EA 56 USD
TS11623-1000 TS11623-5000
Dimethyl glutarate 98%
Dimethyl glutarate
Formula: C₇H₁₂O₄
MW: 160.17 g/mol
Boiling Pt: 211 °C (1013 hPa)
Melting Pt: –37 °C
Density: 1.087 g/cm³
Flash Pt: 103 °C
Storage Temperature: Ambient
MDL Number: MFCD00008468
CAS Number: 1119-40-0
EINECS: 214-277-2
Merck Index: 13,04486

Order Now

Specification Test Results

Appearance Clear colorless Liquid
Infrared spectrum Conforms
Refractive index 1.4236 to 1.4256 (20 °C, 589 nm)
GC ≥97.5 %
Sulfate (SO4) ≤0.005 %
Iron (Fe) ≤5 ppm
Heavy metals (as Pb) ≤5 ppm
Insoluble matter ≤0.005 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR