Dimethyl-3-oxoglutarate 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Dimethyl 1,3-acetonedicarboxylate, Dimethyl acetone-1,3-dicarboxylate

11570-0250 11570-1000 11570-5000
TS11570-0250EA 43 USD
TS11570-0250 TS11570-1000 TS11570-5000
Dimethyl-3-oxoglutarate 97%
Dimethyl-3-oxoglutarate
Formula: C₇H₁₀O₅
Boiling Pt: 237…241 °C (760 mmHg)
Melting Pt: 16…17 °C
Density: 1.18 g/cm³
Flash Pt: >110 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00008462
CAS Number: 1830-54-2
EINECS: 217-385-8

Order Now

Specification Test Results

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4430 to 1.4450 (20 °C, 589 nm)
GC ≥96.0 %
Visual transition interval pH 9.0 (blue)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR