3',5'-Dimethoxy-4'-hydroxyacetophenone 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 3′,5′-Dimethoxy-4′-hydroxyacetophenone, 4'-Hydroxy-3',5'-dimethoxyacetophenone, 1-(3,5-Dimethoxy-4-hydroxyphenyl)ethan-1-one, Acetosyringone, 1-(4-Hydroxy-3,5-dimethoxyphenyl)ethanone

11554-0010 11554-0050 11554-0250
TS11554-0010EA 48 USD
TS11554-0010 TS11554-0050 TS11554-0250
3',5'-Dimethoxy-4'-hydroxyacetophenone 97%
3',5'-Dimethoxy-4'-hydroxyacetophenone
Formula: C₁₀H₁₂O₄
MW: 196.2 g/mol
Melting Pt: 123…128 °C
Storage Temperature: Ambient
MDL Number: MFCD00008748
CAS Number: 2478-38-8
EINECS: 219-610-5

Order Now

Specification Test Results

Appearance Beige to light brown Crystalline powder
Infrared spectrum Conforms
Melting point 123 to 128 °C
GC ≥96.0 %
Titration with NaOH ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR