2,5-Dimethoxybenzaldehyde 97%

Supplier: Thermo Scientific Chemicals
Warning

11539-0250 11539-1000 11539-5000
TS11539-0250EA 33 USD
TS11539-0250 TS11539-1000 TS11539-5000
2,5-Dimethoxybenzaldehyde 97%
2,5-Dimethoxybenzaldehyde
Formula: C₉H₁₀O₃
Boiling Pt: 146 °C (10 mmHg)
Melting Pt: 45…50 °C
Flash Pt: 113 °C
Storage Temperature: Ambient
MDL Number: MFCD00003314
CAS Number: 93-02-7
EINECS: 202-211-5

Order Now

Specification Test Results

Appearance Yellow to beige Crystalline powder or crystals and/or chunks
Infrared spectrum Conforms
Melting point 46 to 51 °C
Water ≤0.5 %
GC ≥96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR