2,4-Dimethoxybenzaldehyde 98%

Supplier: Thermo Scientific Chemicals
Warning

11538-0250 11538-1000 11538-5000
TS11538-0250EA 60 USD
TS11538-0250 TS11538-1000 TS11538-5000
2,4-Dimethoxybenzaldehyde 98%
2,4-Dimethoxybenzaldehyde
Formula: C₉H₁₀O₃
MW: 166.18 g/mol
Boiling Pt: 165 °C (10 mmHg)
Melting Pt: 68…72 °C
Storage Temperature: Ambient
MDL Number: MFCD00003311
CAS Number: 613-45-6
EINECS: 210-342-4

Order Now

Specification Test Results

Appearance White to light brown Crystalline powder
Infrared spectrum Conforms
Melting point 68 to 72 °C
GC ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR