2,3-Dimethoxybenzaldehyde 97%

Supplier: Thermo Scientific Chemicals
Warning

11537-0100 11537-0500
TS11537-0100EA 38 USD
TS11537-0100 TS11537-0500
2,3-Dimethoxybenzaldehyde 97%
2,3-Dimethoxybenzaldehyde
Formula: C₉H₁₀O₃
MW: 166.18 g/mol
Boiling Pt: 137 °C (12 mmHg)
Melting Pt: 49…54 °C
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD00003309
CAS Number: 86-51-1
EINECS: 201-677-7

Order Now

Specification Test Results

Appearance Off-white to beige Crystalline powder or crystals and/or chunks
Infrared spectrum Conforms
Melting point 48 to 53 °C
GC ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR