2',5'-Dimethoxyacetophenone 99%
Supplier: Thermo Scientific Chemicals
|
Warning
|
11534-0050
11534-0250
11534-1000
TS11534-0050EA
27
USD
TS11534-0050
TS11534-0250
TS11534-1000
2',5'-Dimethoxyacetophenone 99%
2',5'-Dimethoxyacetophenone
Formula:
C₁₀H₁₂O₃ Boiling Pt: 155…158 °C (11 mmHg) Melting Pt: 18…20 °C Density: 1.13 g/cm³ Flash Pt: >112 °C Storage Temperature: Ambient |
MDL Number:
MFCD00008728 CAS Number: 1201-38-3 EINECS: 214-858-0 |
Specification Test Results
Appearance | Clear yellow to yellow-green Liquid |
Infrared spectrum | Conforms |
Refractive index | 1.5421 to 1.5441 (20 °C, 589 nm) |
GC | ≥98.5 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...