3,4-Dihydroxybenzaldehyde 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Protocatechuic aldehyde, Protocatechualdehyde

11485-0250 11485-1000
TS11485-0250EA 130 USD
TS11485-0250 TS11485-1000
3,4-Dihydroxybenzaldehyde 97%
3,4-Dihydroxybenzaldehyde
Formula: C₇H₆O₃
MW: 138.12 g/mol
Boiling Pt: 295 °C (1013 hPa)
Melting Pt: 152…157 °C
Storage Temperature: Ambient
MDL Number: MFCD00003370
CAS Number: 139-85-5
EINECS: 205-377-7
Merck Index: 12,08079

Order Now

Specification Test Results

Appearance Beige to grey-brown Crystalline powder
Infrared spectrum Conforms
Melting point 152 to 157 °C
HPLC ≥96.0 %
Copper (Cu) ≤50 ppm
Iron (Fe) ≤100 ppm
Lead (Pb) ≤50 ppm

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR