2,5-Dihydroxybenzaldehyde 99%

Supplier: Thermo Scientific Chemicals
Warning

11484-0050 11484-0250
TS11484-0050EA 95 USD
TS11484-0050 TS11484-0250
2,5-Dihydroxybenzaldehyde 99%
2,5-Dihydroxybenzaldehyde
Formula: C₇H₆O₃
MW: 138.12 g/mol
Melting Pt: 98…102 °C
Storage Temperature: Ambient
MDL Number: MFCD00003333
CAS Number: 1194-98-5
EINECS: 214-789-6

Order Now

Specification Test Results

Appearance Yellow to khaki Crystalline powder
Infrared spectrum Conforms
Melting point 98 to 102 °C
GC ≥98.5 %
Copper (Cu) ≤50 ppm
Iron (Fe) ≤100 ppm
Lead (Pb) ≤50 ppm

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR