1,8-Dihydroxy-9,10-anthraquinone 95%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Dantron, Danthron, 1,8-Dihydroxyanthraquinone

11483-0050 11483-1000 11483-5000
TS11483-0050EA 32 USD
TS11483-0050 TS11483-1000 TS11483-5000
1,8-Dihydroxy-9,10-anthraquinone 95%
1,8-Dihydroxy-9,10-anthraquinone
Formula: C₁₄H₈O₄
MW: 240.22 g/mol
Melting Pt: 190…195 °C
Flash Pt: >200 °C
MDL Number: MFCD00001211
CAS Number: 117-10-2
EINECS: 204-173-5
Merck Index: 13,02841

Order Now

Specification Test Results

Appearance Yellow-green to orange to brown Powder
Infrared spectrum Conforms
Melting point 190 to 195 °C
HPLC ≥94 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR