Diethyl mesoxalate (diethyl ketomalonate) 95%
Supplier: Thermo Scientific Chemicals
|
Warning
|
11443-0050
11443-0250
TS11443-0050EA
48
USD
TS11443-0050
TS11443-0250
Diethyl mesoxalate (diethyl ketomalonate) 95%
Diethyl mesoxalate (diethyl ketomalonate)
Formula:
C₇H₁₀O₅ Boiling Pt: 208…210 °C (760 mmHg) Density: 1.142 g/cm³ Flash Pt: >112 °C Storage Temperature: Ambient |
MDL Number:
MFCD00009121 CAS Number: 609-09-6 EINECS: 210-176-2 |
Specification Test Results
Appearance | Clear yellow Liquid |
Infrared spectrum | Conforms |
Refractive index | 1.4150 to 1.4190 (20 °C, 589 nm) |
GC | ≥94 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...