(±)-Diethyl acetylsuccinate 99%

Supplier: Thermo Scientific Chemicals

11395-0250 11395-1000 11395-5000
TS11395-0250EA 61 USD
TS11395-0250 TS11395-1000 TS11395-5000
(±)-Diethyl acetylsuccinate 99%
(±)-Diethyl acetylsuccinate
Formula: C₁₀H₁₆O₅
Boiling Pt: 180…183 °C (50 mmHg)
Melting Pt: –8 °C
Density: 1.08 g/cm³
Flash Pt: >112 °C
Storage Temperature: Ambient
MDL Number: MFCD00009157
CAS Number: 1115-30-6
EINECS: 214-223-8

Order Now

Specification Test Results

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4340 to 1.4360 (20 °C, 589 nm)
GC ≥98.5 %
Chloride (Cl) ≤20 ppm
Nitrite (NO2) ≤0.5 ppm
Sulfate (SO4) ≤100 ppm
Insoluble matter ≤0.02 % (in sodium carbonate solution)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR