2,6-Dichlorobenzaldehyde 99%

Supplier: Thermo Scientific Chemicals
Danger

11316-0010 11316-0500 11316-2500
TS11316-0010EA 537 USD
TS11316-0010 TS11316-0500 TS11316-2500
2,6-Dichlorobenzaldehyde 99%
2,6-Dichlorobenzaldehyde
Formula: C₇H₄Cl₂O
Boiling Pt: 239 °C (1013 hPa)
Melting Pt: 71…72 °C
Flash Pt: >100 °C
Storage Temperature: Ambient
MDL Number: MFCD00003307
CAS Number: 83-38-5
EINECS: 201-472-2
UN: 3261
ADR: 8,III

Order Now

Specification Test Results

Appearance White to light beige Crystalline powder
Infrared spectrum Conforms
Melting point 68 to 71 °C
GC ≥98.5 %
Specific optical rotation -33° to -39° (20 °C, 589 nm) (c=1, CHCl3)
Titration with NaOH ≥98.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR