ω,4-Dibromoacetophenone 98%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Tau Protein Kinase I Inhibitor, 2,4'-Dibromoacetophenone, Glycogen Synthase Kinase-3β Inhibitor VII, α,4-Dibromoacetophenone, GSK-3β Inhibitor VII, TPK I Inhibitor, 2-Bromo-1-(4-bromophenyl)-1-ethanone, 4-Bromophenacyl bromide

11268-0100 11268-0500 11268-2500
TS11268-0100EA 51 USD
TS11268-0100 TS11268-0500 TS11268-2500
ω,4-Dibromoacetophenone 98%
ω,4-Dibromoacetophenone
Formula: C₈H₆Br₂O
MW: 277.94 g/mol
Boiling Pt: 1415 °C (760 mmHg)
Melting Pt: 107…111 °C
Storage Temperature: Ambient
MDL Number: MFCD00000200
CAS Number: 99-73-0
EINECS: 202-783-6
UN: 3261
ADR: 8,II
Merck Index: 13,01412

Order Now

Specification Test Results

Appearance Light yellow to beige Crystalline solid
Infrared spectrum Conforms
Melting point 107 to 111 °C
GC ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR