Deoxybenzoin 98%

Supplier: Thermo Scientific Chemicals

11196-0250 11196-1000
TS11196-0250EA 51 USD
TS11196-0250 TS11196-1000
Deoxybenzoin 98%
Deoxybenzoin
Formula: C₁₄H₁₂O
Melting Pt: 52…57 °C
Flash Pt: >110 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00003081
CAS Number: 451-40-1
EINECS: 207-193-2

Order Now

Specification Test Results

Appearance White to yellow Crystalline powder or crystals and/or chunks
Infrared spectrum Conforms
Melting point 52 to 57 °C
GC ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR