p-Cymene 99+%

Supplier: Thermo Scientific Chemicals
Danger

11176-0010 11176-0025 11176-0100 11176-2500
TS11176-0010EA 72 USD
TS11176-0010 TS11176-0025 TS11176-0100 TS11176-2500
p-Cymene 99+%
p-Cymene
Formula: C₁₀H₁₄
Boiling Pt: 176…178 °C (760 mmHg)
Melting Pt: –68 °C
Density: 0.854 g/cm³
Flash Pt: 47 °C
Storage Temperature: Ambient
MDL Number: MFCD00008893
CAS Number: 99-87-6
EINECS: 202-796-7
UN: 2046
ADR: 3,III
Merck Index: 13,02792

Order Now

Specification Test Results

Appearance Clear liquid
Infrared spectrum Conforms
Refractive index 1.4890 to 1.4910 (20 °C, 589 nm)
GC ≥99.0 %
Color scale ≤30 APHA
Specific gravity (25°C) 0.853 to 0.860


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR