Cyclopropyl methyl ketone 98%
Supplier: Thermo Scientific Chemicals
|
Danger
|
11171-0250
11171-1000
11171-5000
TS11171-0250EA
51
USD
TS11171-0250
TS11171-1000
TS11171-5000
Cyclopropyl methyl ketone 98%
Cyclopropyl methyl ketone
Formula:
C₅H₈O Boiling Pt: 111 °C (760 mmHg) Melting Pt: –68 °C Density: 0.898 g/cm³ Flash Pt: 21 °C Storage Temperature: Ambient |
MDL Number:
MFCD00001297 CAS Number: 765-43-5 EINECS: 212-146-4 UN: 2924 ADR: 3,II |
Specification Test Results
Appearance | Clear colorless to light yellow Liquid |
Infrared spectrum | Conforms |
Refractive index | 1.4220 to 1.4260 (20 °C, 589 nm) |
Water | ≤0.2 % |
GC | ≥97.5 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...