Cyclopentanol 99%
Supplier: Thermo Scientific Chemicals
|
Warning
|
11152-1000
11152-5000
TS11152-1000EA
37
USD
TS11152-1000
TS11152-5000
Cyclopentanol 99%
Cyclopentanol
Formula:
C₅H₁₀O MW: 86.13 g/mol Boiling Pt: 139 °C (1013 hPa) Melting Pt: –19 °C Density: 0.94 g/cm³ Flash Pt: 51 °C Storage Temperature: Ambient |
MDL Number:
MFCD00001363 CAS Number: 96-41-3 EINECS: 202-504-8 UN: 2244 ADR: 3,III Merck Index: 13,02770 |
Specification Test Results
Appearance | Clear colorless to light yellow Liquid |
Infrared spectrum | Conforms |
Refractive index | 1.4520 to 1.4540 (20 °C, 589 nm) |
GC | ≥98.5 % |
Color scale | ≤50 APHA |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...