Cyclohexylméthanol 99%

Supplier: Thermo Scientific Chemicals

11134-0050 11134-0250 11134-1000
TS11134-0050EA 25 USD
TS11134-0050 TS11134-0250 TS11134-1000
Cyclohexylméthanol 99%
Cyclohexylméthanol
Formula: C₇H₁₄O
Boiling Pt: 181 °C (760 mmHg)
Density: 0.91 g/cm³
Flash Pt: 71 °C
Storage Temperature: Ambient
MDL Number: MFCD00001510
CAS Number: 100-49-2
EINECS: 202-857-8
Merck Index: 13,02760

Order Now

Specification Test Results

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
GC ≥98.5 %
HPLC ≥96.0 %
Specific optical rotation -105° to -109° (20°C, 589 nm) (C=1, methanol)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR