(±)-β-Citronellol 95%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: DL-Citronellol, (±)-Citronellol, Citronellol, β-Citronellol, 3,7-Dimethyloct-6-en-1-ol

11046-0050 11046-1000 11046-5000
TS11046-0050EA 35 USD
TS11046-0050 TS11046-1000 TS11046-5000
(±)-β-Citronellol 95%
(±)-β-Citronellol
Formula: C₁₀H₂₀O
MW: 156.27 g/mol
Boiling Pt: 222 °C (1013 hPa)
Density: 0.854 g/cm³
Flash Pt: 79 °C
MDL Number: MFCD00002935
CAS Number: 106-22-9
EINECS: 203-375-0
Merck Index: 13,02354

Order Now

Specification Test Results

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4540 to 1.4580 (20 °C, 589 nm)
GC ≥94 %
Specific gravity (25°C) 0.850 to 0.860

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR