Citraconic acid 99+%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 2-Methyl-2-butenedioic acid

11043-0050 11043-0250 11043-1000
TS11043-0050EA 28 USD
TS11043-0050 TS11043-0250 TS11043-1000
Citraconic acid 99+%
Citraconic acid
Formula: C₅H₆O₄
MW: 130.1 g/mol
Melting Pt: 88…92 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00078086
CAS Number: 498-23-7
EINECS: 207-858-7
Merck Index: 13,02345

Order Now

Specification Test Results

Appearance White to light beige Crystalline powder
Infrared spectrum Conforms
Titration with NaOH ≥99.0 %
UV Lambda max 630 to 635 nm (in water)
Absorption Ratio P-15/P+15 0.86 to 1.11
Visual transition interval from pH 0.2 (yellow) to pH 1.8 (blue)
BSC Certification Passes

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR