1-Chloro-2-nitrobenzene 99+%
Supplier: Thermo Scientific Chemicals
|
Danger
|
10962-0010
10962-1000
TS10962-0010EA
69
USD
TS10962-0010
TS10962-1000
1-Chloro-2-nitrobenzene 99+%
1-Chloro-2-nitrobenzene
Formula:
C₆H₄ClNO₂ Boiling Pt: 246 °C (1013 hPa) Melting Pt: 32…34 °C Density: 1.348 g/cm³ Flash Pt: 124 °C Storage Temperature: Ambient |
MDL Number:
MFCD00007061 CAS Number: 88-73-3 EINECS: 201-854-9 UN: 1578 ADR: 6.1,II Merck Index: 13,02170 |
Specification Test Results
Appearance | Yellow Fused crystalline mass |
Infrared spectrum | Conforms |
Melting point | 32 to 34 °C |
GC | ≥99.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...