3-Chloro-1-propanol 98% stabilized
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
3-Chloropropan-1-ol, 1-Chloro-3-hydroxypropane
10929-0250
10929-1000
10929-5000
TS10929-0250EA
33
USD
TS10929-0250
TS10929-1000
TS10929-5000
3-Chloro-1-propanol 98% stabilized
3-Chloro-1-propanol
Formula:
C₃H₇ClO MW: 94.54 g/mol Boiling Pt: 160…162 °C (760 mmHg) Melting Pt: –20 °C Density: 1.13 g/cm³ Flash Pt: 75 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00002943 CAS Number: 627-30-5 EINECS: 210-992-9 UN: 2849 ADR: 6.1,III |
Specification Test Results
Appearance | Clear colorless to yellow Liquid |
Infrared spectrum | Conforms |
Refractive index | 1.4450 to 1.4500 (20 °C, 589 nm) |
GC | ≥97.5 % |
Stabilizer | 0.1 % Na2CO3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...