2-Chlorocyclohexanone 95% stabilised

Supplier: Thermo Scientific Chemicals
Warning

10896-0050 10896-0250 10896-1000
TS10896-0050EA 32 USD
TS10896-0050 TS10896-0250 TS10896-1000
2-Chlorocyclohexanone 95% stabilised
2-Chlorocyclohexanone
Formula: C₆H₉ClO
Boiling Pt: 82…83 °C (10 mmHg)
Melting Pt: 23 °C
Density: 1.16 g/cm³
Flash Pt: 82 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00001626
CAS Number: 822-87-7
EINECS: 212-505-5

Order Now

Specification Test Results

Appearance Clear brown Liquid
Infrared spectrum Conforms
Refractive index 1.4830 to 1.4850 (20 °C, 589 nm)
GC ≥94 %
Stabilizer 0.2 % CaCO3/MgO (1:1)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR