4'-Chloroacetophenone 98+%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: p-Chloroacetophenone

10854-0010 10854-0050 10854-2500
TS10854-0010EA 211 USD
TS10854-0010 TS10854-0050 TS10854-2500
4'-Chloroacetophenone 98+%
4'-Chloroacetophenone
Formula: C₈H₇ClO
MW: 154.6 g/mol
Boiling Pt: 232 °C (760 mmHg)
Melting Pt: 18 °C
Density: 1.19 g/cm³
Flash Pt: 90 °C
Storage Temperature: Ambient
MDL Number: MFCD00000624
CAS Number: 99-91-2
EINECS: 202-800-7
Merck Index: 13,02133

Order Now

Specification Test Results

Appearance Clear colorless to yellow Liquid after melting
Infrared spectrum Conforms
Refractive index 1.5539 to 1.5559 (20 °C, 589 nm)
GC ≥96.0 %
Specific optical rotation -7° to -8.5° (20°C, 589 nm) (c=1.13, MeOH)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR