Chloroacetaldehyde dimethyl acetal 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 1-Chloro-2,2-dimethoxyethane

10850-0010 10850-2500
TS10850-0010EA 100 USD
TS10850-0010 TS10850-2500
Chloroacetaldehyde dimethyl acetal 97%
Chloroacetaldehyde dimethyl acetal
Formula: C₄H₉ClO₂
Boiling Pt: 128 °C (1013 hPa)
Melting Pt: –73 °C
Density: 1.09 g/cm³
Flash Pt: 28 °C
Storage Temperature: Ambient
MDL Number: MFCD00000948
CAS Number: 97-97-2
EINECS: 202-624-0
UN: 1993
ADR: 3,III

Order Now

Specification Test Results

Appearance Clear colorless to yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4140 to 1.4160 (20 °C, 589 nm)
GC ≥96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR