Methyl-4-chloro-4-oxobutanoate 97%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Methyl succinyl chloride, Succinic acid monomethyl ester chloride, Methyl 4-Chloro-4-oxobutyrate, Methyl Succinyl chloride, 4-Chloro-4-oxobutyric acid methyll ester

10840-0050 10840-0250 10840-1000
TS10840-0050EA 46 USD
TS10840-0050 TS10840-0250 TS10840-1000
Methyl-4-chloro-4-oxobutanoate 97%
Methyl-4-chloro-4-oxobutanoate
Formula: C₅H₇ClO₃
MW: 150.56 g/mol
Boiling Pt: 58…65 °C (3 mmHg)
Density: 1.232 g/cm³
Flash Pt: 73 °C
Storage Temperature: Ambient
MDL Number: MFCD00000750
CAS Number: 1490-25-1
UN: 3265
ADR: 8 II

Order Now

Specification Test Results

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4390 to 1.4410 (20 °C, 589 nm)
GC ≥96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR