Butyrylthiocholine iodide 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: S-n-Butyrylthiocholine iodide, (2-Butyrylthioethyl)trimethylammonium iodide, 2-(Butyrylthio)-N,N,N-trimethylethan-1-aminium iodide, (Propylcarbonylthioethyl)trimethylammonium iodide, (2-(Butyrylthio)ethyl)trimethylammonium iodide

10815-0250 10815-1000
TS10815-0250EA 148 USD
TS10815-0250 TS10815-1000
Butyrylthiocholine iodide 98%
Butyrylthiocholine iodide
Formula: H₃C(CH₂)₂C(S)O(CH₂)₂N(CH₃)₃I
Melting Pt: 172…176 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00011845
CAS Number: 1866-16-6
EINECS: 217-475-7

Order Now

Specification Test Results

Appearance White Crystalline powder or crystals
Infrared spectrum Conforms
Melting point 172 to 176 °C
Titration Argentometric ≥97.5 %
UV at 250 nm A: ≤0.01
Color scale ≤10 APHA
Residue after evaporation ≤0.0005 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR