tert-Butyl acetoacetate 97%
Supplier: Thermo Scientific Chemicals
Synonyms:
tert-Butyl Acetoacetate
10778-1000
10778-5000
TS10778-1000EA
46
USD
TS10778-1000
TS10778-5000
tert-Butyl acetoacetate 97%
tert-Butyl acetoacetate
Formula:
C₈H₁₄O₃ Boiling Pt: 190 °C (1013 hPa) Melting Pt: –38 °C Density: 0.97 g/cm³ Flash Pt: 76 °C |
MDL Number:
MFCD00008811 CAS Number: 1694-31-1 EINECS: 216-904-5 |
Specification Test Results
Appearance | Clear colorless to yellow Liquid |
Infrared spectrum | Conforms |
Refractive index | 1.4180 to 1.4210 (20 °C, 589 nm) |
GC | ≥96.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...